APO455830476 Apollo Scientific Ltd

4-(Hydroxymethyl)-2-methyl-1H-imidazole

  • MDL number MFCD06202901
  • Cas Number : 45533-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Synonym (2-Methyl-1H-imidazol-4-yl)methanol
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=35955

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=35955
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart