APO455830478 Apollo Scientific Ltd

6-Amino-2-methyl-2H-indazole

  • MDL number MFCD00464866
  • Cas Number : 50593-30-1
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 148-150
Iupac Name 2-methyl-2H-indazol-6-ylamine
Inchi Code 1S/C8H9N3/c1-11-5-6-2-3-7(9)4-8(6)10-11/h2-5H,9H2,1H3
Inchi Key MHCWLERQNFATHZ-UHFFFAOYSA-N

Description

Synonym 2-Methyl-2H-indazol-6-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 148-150
Product Link https://store.apolloscientific.co.uk/product/6-amino-2-methyl-2h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-amino-2-methyl-2h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart