APO455830479 Apollo Scientific Ltd

4-(4-Methoxyphenyl)pyrimidin-2-amine

  • MDL number MFCD00665912
  • Cas Number : 99844-02-7
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-methoxyphenyl)-2-pyrimidinamine
Inchi Code 1S/C11H11N3O/c1-15-9-4-2-8(3-5-9)10-6-7-13-11(12)14-10/h2-7H,1H3,(H2,12,13,14)
Inchi Key GHSMJZASRNAUBJ-UHFFFAOYSA-N

Description

Synonym 2-Amino-4-(4-methoxyphenyl)pyrimidine, 4-(2-Aminopyrimidin-4-yl)anisole
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-4-methoxyphenylpyrimidin-2-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methoxyphenylpyrimidin-2-amine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart