APO455830480 Apollo Scientific Ltd

6-Amino-1-methyl-1H-indazole

  • MDL number MFCD00456076
  • Cas Number : 74728-65-7
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 174-176
Iupac Name 1-methyl-1H-indazol-6-amine
Inchi Code 1S/C8H9N3/c1-11-8-4-7(9)3-2-6(8)5-10-11/h2-5H,9H2,1H3
Inchi Key YTKNUPJYGSOVLV-UHFFFAOYSA-N

Description

Synonym 1-Methyl-1H-indazol-6-amine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 174-176
Product Link https://store.apolloscientific.co.uk/product/6-amino-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-amino-1-methyl-1h-indazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart