APO455830481 Apollo Scientific Ltd

1-Methyl-4-nitro-1H-indazole

  • MDL number MFCD09607929
  • Cas Number : 26120-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-methyl-4-nitro-1h-indazole

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/1-methyl-4-nitro-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart