APO455830485 Apollo Scientific Ltd

2-Bromo-5-chloropyridine-4-boronic acid

  • MDL number MFCD08064041
  • Cas Number : 1072952-51-2
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-2-bromo-5-chloropyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-chloropyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P260;P280;P301+P312;P302+P352;P304+P340;P361+P364
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-chloropyridine-4-boronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart