APO455830491 Apollo Scientific Ltd

3-Amino-2-phenylpyridine

  • MDL number MFCD04114113
  • Cas Number : 101601-80-3
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 62-64
Iupac Name 2-phenyl-3-pyridinylamine
Inchi Code 1S/C11H10N2/c12-10-7-4-8-13-11(10)9-5-2-1-3-6-9/h1-8H,12H2
Inchi Key XTHJCITVHCRQRD-UHFFFAOYSA-N

Description

Synonym 2-Phenylpyridin-3-amine, (3-Aminopyridin-2-yl)benzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 62-64
Product Link https://store.apolloscientific.co.uk/product/3-amino-2-phenylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-2-phenylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart