APO455830492 Apollo Scientific Ltd

3-Amino-4-hydroxypyridine

  • MDL number MFCD04114230
  • Cas Number : 6320-39-4
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 145-149
Iupac Name 3-amino-4-pyridinol
Inchi Code 1S/C5H6N2O/c6-4-3-7-2-1-5(4)8/h1-3H,6H2,(H,7,8)
Inchi Key OIIBRAGQGFLUFI-UHFFFAOYSA-N

Description

Synonym 3-Aminopyridin-4-ol, 4-Hydroxypyridin-3-amine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 145-149
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-hydroxypyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart