APO455830500 Apollo Scientific Ltd

2-Phenylnicotinic acid

  • MDL number MFCD03069417
  • Cas Number : 33421-39-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Phenylpyridine-3-carboxylic acid, 3-Carboxy-2-phenylpyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-phenylnicotinic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenylnicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart