APO455830512 Apollo Scientific Ltd

Ethyl 3-bromo-5-nitrobenzoate

  • MDL number MFCD09258760
  • Cas Number : 690260-94-7
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromo-5-nitrobenzoate
Inchi Code 1S/C9H8BrNO4/c1-2-15-9(12)6-3-7(10)5-8(4-6)11(13)14/h3-5H,2H2,1H3
Inchi Key PUBVWGDKFHIPRR-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-bromo-5-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-bromo-5-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart