APO455830514 Apollo Scientific Ltd

N,N-Bis(isopropyl)benzylamine

  • MDL number MFCD07368226
  • Cas Number : 34636-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Benzyl-N-(prop-2-yl)propan-2-amine
Density 0.9
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36138

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36138
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart