APO455830517 Apollo Scientific Ltd

4-(3-Bromobenzyl)morpholine

  • MDL number MFCD00979456
  • Cas Number : 364793-82-8
  • Molecular Weight : 256.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(3-Bromophenyl)methyl]morpholine
Density 1.40
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-bromobenzylmorpholine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-bromobenzylmorpholine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart