APO455830520 Apollo Scientific Ltd

6-Chloro-4-methylpyridine-3-boronic acid

  • MDL number MFCD07368882
  • Cas Number : 913836-08-5
  • Molecular Weight : 171.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 105-107
Iupac Name 6-chloro-4-methyl-3-pyridinylboronic acid
Inchi Code 1S/C6H7BClNO2/c1-4-2-6(8)9-3-5(4)7(10)11/h2-3,10-11H,1H3
Inchi Key HMDMMWUUIMJRAQ-UHFFFAOYSA-N

Description

Synonym 5-Borono-2-chloro-4-methylpyridine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 105-107
Product Link https://store.apolloscientific.co.uk/product/6-chloro-4-methylpyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-4-methylpyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart