APO455830528 Apollo Scientific Ltd

6-Carboxypyridine-3-boronic acid

  • MDL number MFCD09027238
  • Cas Number : 913836-11-0
  • Molecular Weight : 166.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 5-Boronopicolinic acid, 5-Boronopyridine-2-carboxylic acid
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-carboxypyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-carboxypyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart