APO455830530 Apollo Scientific Ltd

4-[(3-Chloro-4-methylphenyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD09027211
  • Cas Number : 913835-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-219
Synonym N-(3-Chloro-4-methylphenyl) 4-boronobenzamide
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36025

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36025
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart