APO455830535 Apollo Scientific Ltd

4-[(2,3-Dimethylphenyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD09027210
  • Cas Number : 913835-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-N-(2,3-dimethylphenyl)benzamide
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-23-dimethylphenylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-23-dimethylphenylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart