APO455830536 Apollo Scientific Ltd

4-[(2,4-Dimethylphenyl)carbamoyl]benzeneboronic acid

  • MDL number MFCD09027212
  • Cas Number : 913835-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-N-(2,4-dimethylphenyl)benzamide
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36091

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36091
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart