APO455830542 Apollo Scientific Ltd

3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde

  • MDL number MFCD07772845
  • Cas Number : 81068-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[2-(3-Formylphenoxy)ethyl]morpholine
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid-Viscous
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31478

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31478
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart