APO455830551 Apollo Scientific Ltd

5-Acetyl-2-amino-4-methylpyrimidine

  • MDL number MFCD00052621
  • Cas Number : 66373-25-9
  • Molecular Weight : 151.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 226-227
Iupac Name 1-(2-amino-4-methyl-5-pyrimidinyl)ethanone
Inchi Code 1S/C7H9N3O/c1-4-6(5(2)11)3-9-7(8)10-4/h3H,1-2H3,(H2,8,9,10)
Inchi Key LWZQQJIAMSDELT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 226-227
Product Link https://store.apolloscientific.co.uk/product/5-acetyl-2-amino-4-methylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-acetyl-2-amino-4-methylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart