APO455830560 Apollo Scientific Ltd

6-[4-(tert-Butoxycarbonyl)piperazin-1-yl]nicotinic acid

  • MDL number MFCD06245543
  • Cas Number : 201809-22-5
  • Molecular Weight : 307.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 298-300
Synonym 6-[4-(tert-Butoxycarbonyl)piperazin-1-yl]pyridine-3-carboxylic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-4-tert-butoxycarbonylpiperazin-1-ylnicotinic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-4-tert-butoxycarbonylpiperazin-1-ylnicotinic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart