APO455830570 Apollo Scientific Ltd

1-(2-Chlorophenyl)biguanide hydrochloride

  • MDL number MFCD00053016
  • Cas Number : 19579-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-134
Synonym N-(2-Chlorophenyl)imidodicarbonimidic diamide hydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13871

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13871
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart