APO455830582 Apollo Scientific Ltd

4-(2-Cyanoprop-2-yl)benzeneboronic acid

  • MDL number MFCD06659832
  • Cas Number : 850568-67-1
  • Molecular Weight : 189.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 146-148
Iupac Name 4-(1-cyano-1-methylethyl)phenylboronic acid
Inchi Code 1S/C10H12BNO2/c1-10(2,7-12)8-3-5-9(6-4-8)11(13)14/h3-6,13-14H,1-2H3
Inchi Key BNXBFDTWYHAEIR-UHFFFAOYSA-N

Description

Synonym 2-(4-Boronophenyl)-2-methylpropanenitrile, 2-(4-Boronophenyl)-2-methylpropionitrile
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 146-148
Product Link https://store.apolloscientific.co.uk/product/4-2-cyanoprop-2-ylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-2-cyanoprop-2-ylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart