APO455830585 Apollo Scientific Ltd

4-(Isopropylsulphonyl)benzeneboronic acid

  • MDL number MFCD06659866
  • Cas Number : 850567-98-5
  • Molecular Weight : 228.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 126-128
Iupac Name 4-(isopropylsulfonyl)phenylboronic acid
Inchi Code 1S/C9H13BO4S/c1-7(2)15(13,14)9-5-3-8(4-6-9)10(11)12/h3-7,11-12H,1-2H3
Inchi Key IGJLXIUSOMMUIV-UHFFFAOYSA-N

Description

Synonym 4-[(Prop-2-yl)sulphonyl]benzeneboronic acid, 4-Boronophenyl isopropyl sulphone
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126-128
Product Link https://store.apolloscientific.co.uk/product/4-isopropylsulphonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-isopropylsulphonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart