APO455830590 Apollo Scientific Ltd

3-(Diethylcarbamoyl)benzeneboronic acid

  • MDL number MFCD04115684
  • Cas Number : 237413-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-136
Synonym 3-Borono-N,N-diethylbenzamide, 3-[(Diethylamino)carbonyl]benzeneboronic acid
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-diethylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-diethylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart