APO455830596 Apollo Scientific Ltd

3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]benzoic acid

  • MDL number MFCD00203067
  • Cas Number : 387350-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-208
Synonym 2-(3-Carboxyphenoxy)-4,6-dimethoxypyrimidine, 2-(3-Carboxyphenoxy)-4,6-dimethoxy-1,3-diazine
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14410

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14410
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart