APO455830597 Apollo Scientific Ltd

3-(Piperidine-1-carbonyl)benzeneboronic acid

  • MDL number MFCD04115686
  • Cas Number : 850568-34-2
  • Molecular Weight : 233.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 150-154
Iupac Name 3-(1-piperidinylcarbonyl)phenylboronic acid
Inchi Code 1S/C12H16BNO3/c15-12(14-7-2-1-3-8-14)10-5-4-6-11(9-10)13(16)17/h4-6,9,16-17H,1-3,7-8H2
Inchi Key XFTARKGZGNWJJT-UHFFFAOYSA-N

Description

Synonym N-Piperidinyl 3-boronobenzamide
Purity 97%
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 150-154
Product Link https://store.apolloscientific.co.uk/product/3-piperidine-1-carbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-piperidine-1-carbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart