APO455830600 Apollo Scientific Ltd

3-(3,4-Dimethoxyphenyl)-1H-pyrazole

  • MDL number MFCD02091527
  • Cas Number : 154257-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Dimethoxy-4-(1H-pyrazol-3-yl)benzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14401

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14401
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart