APO455830603 Apollo Scientific Ltd

5-Methoxyisatoic anhydride

  • MDL number MFCD02179411
  • Cas Number : 37795-77-0
  • Molecular Weight : 193.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244-246
Synonym 6-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione, 6-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methoxyisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxyisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart