APO455830605 Apollo Scientific Ltd

4-Nitroisatoic anhydride

  • MDL number MFCD00798220
  • Cas Number : 63480-10-4
  • Molecular Weight : 208.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 253
Iupac Name 7-nitro-2H-3,1-benzoxazine-2,4(1H)-dione
Inchi Code 1S/C8H4N2O5/c11-7-5-2-1-4(10(13)14)3-6(5)9-8(12)15-7/h1-3H,(H,9,12)
Inchi Key KCNIWFFVWBXWAV-UHFFFAOYSA-N

Description

Synonym 7-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione, 7-Nitro-1H-benzo[d][1,3]oxazine-2,4-dione
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 253
Product Link https://store.apolloscientific.co.uk/product/4-nitroisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-nitroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart