APO455830607 Apollo Scientific Ltd

3-Bromoisatoic anhydride

  • MDL number MFCD06738735
  • Cas Number : 331646-98-1
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-2H-3,1-benzoxazine-2,4(1H)-dione
Inchi Code 1S/C8H4BrNO3/c9-5-3-1-2-4-6(5)10-8(12)13-7(4)11/h1-3H,(H,10,12)
Inchi Key YRJWMRDVBYWGKK-UHFFFAOYSA-N

Description

Synonym 8-Bromo-2H-3,1-benzoxazine-2,4(1H)-dione, 8-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromoisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromoisatoic-anhydride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart