APO455830611 Apollo Scientific Ltd

5,7-Dichlorobenzo[b]furan-3(2H)-one

  • MDL number MFCD08544411
  • Cas Number : 74815-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5,7-Dichloro-2,3-dihydro-3-oxobenzo[b]furan, 5,7-Dichloro-1-benzofuran-3(2H)-one
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45465

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45465
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart