APO455830613 Apollo Scientific Ltd

6-Bromobenzo[b]furan-3(2H)-one

  • MDL number MFCD08437617
  • Cas Number : 201809-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-1-benzofuran-3(2H)-one, 6-Bromo-2,3-dihydro-3-oxobenzo[b]furan
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45468

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=45468
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart