APO455830616 Apollo Scientific Ltd

3-(Methylsulphonyl)benzaldehyde

  • MDL number MFCD11052329
  • Cas Number : 43114-43-8
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 88.7-89
Iupac Name 3-(methylsulfonyl)benzaldehyde
Inchi Code 1S/C8H8O3S/c1-12(10,11)8-4-2-3-7(5-8)6-9/h2-6H,1H3
Inchi Key OVSMURBVBQUOSF-UHFFFAOYSA-N

Description

Synonym 3-Formylphenyl methyl sulphone
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 88.7-89
Product Link https://store.apolloscientific.co.uk/product/3-methylsulphonylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylsulphonylbenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart