APO455830622 Apollo Scientific Ltd

4-(Bromomethyl)-2-chlorobenzoic acid

  • MDL number MFCD18378710
  • Cas Number : 290820-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-Bromo-2-chloro-p-toluic acid, 4-Carboxy-3-chlorobenzyl bromide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64220

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=64220
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart