APO455830627 Apollo Scientific Ltd

2-Chloro-5-methylpyrazine

  • MDL number MFCD00055039
  • Cas Number : 59303-10-5
  • Molecular Weight : 128.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-methylpyrazine
Inchi Code 1S/C5H5ClN2/c1-4-2-8-5(6)3-7-4/h2-3H,1H3
Inchi Key XSDAJECCWPYVGW-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-methyl-1,4-diazine
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-methylpyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-methylpyrazine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart