APO455830633 Apollo Scientific Ltd

4-(Pent-1-yl)benzeneboronic acid

  • MDL number MFCD00995151
  • Cas Number : 121219-12-3
  • Molecular Weight : 192.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 82-84
Iupac Name 4-pentylphenylboronic acid
Inchi Code 1S/C11H17BO2/c1-2-3-4-5-10-6-8-11(9-7-10)12(13)14/h6-9,13-14H,2-5H2,1H3
Inchi Key UZRMPSOGFATLJE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 82-84
Product Link https://store.apolloscientific.co.uk/product/4-pent-1-ylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-pent-1-ylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart