APO455830636 Apollo Scientific Ltd

3,5-Dichloropyrazine-2-carboxylic acid

  • MDL number MFCD09909819
  • Cas Number : 312736-49-5
  • Molecular Weight : 192.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-3,5-dichloropyrazine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichloropyrazine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloropyrazine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart