APO455830641 Apollo Scientific Ltd

5-Acetylthiophene-2-carboxaldehyde

  • MDL number MFCD00509280
  • Cas Number : 4565-29-1
  • Molecular Weight : 154.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Acetyl-5-formylthiophene, 2-Ethanoyl-5-formylthiophene, 5-Acetyl-2-thenaldehyde
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-acetylthiophene-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-acetylthiophene-2-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart