APO455830649 Apollo Scientific Ltd

2-Ethynylnaphthalene

  • MDL number MFCD00870424
  • Cas Number : 2949-26-0
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 42
Iupac Name 2-ethynylnaphthalene
Inchi Code 1S/C12H8/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h1,3-9H
Inchi Key IZXPFTLEVNQLGD-UHFFFAOYSA-N

Description

Purity 98%
Flash Point 110
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42
Product Link https://store.apolloscientific.co.uk/product/2-ethynylnaphthalene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-ethynylnaphthalene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart