APO455830655 Apollo Scientific Ltd

3-(2-Aminoethyl)thiophene hydrochloride

  • MDL number MFCD09265521
  • Cas Number : 34843-84-0
  • Molecular Weight : 163.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 214-216
Iupac Name 2-(3-thienyl)ethanamine hydrochloride
Inchi Code 1S/C6H9NS.ClH/c7-3-1-6-2-4-8-5-6;/h2,4-5H,1,3,7H2;1H
Inchi Key PYUCPHDVJORSMA-UHFFFAOYSA-N

Description

Synonym 2-(Thien-3-yl)ethylamine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 214-216
Product Link https://store.apolloscientific.co.uk/product/3-2-aminoethylthiophene-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-2-aminoethylthiophene-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart