APO455830656 Apollo Scientific Ltd

5-Bromo-1H-benzimidazole-2-carboxylic acid

  • MDL number MFCD06738769
  • Cas Number : 40197-20-4
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 158
Iupac Name 5-bromo-1H-benzimidazole-2-carboxylic acid
Inchi Code 1S/C8H5BrN2O2/c9-4-1-2-5-6(3-4)11-7(10-5)8(12)13/h1-3H,(H,10,11)(H,12,13)
Inchi Key ULTDLMITPWHISY-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-carboxy-1H-benzimidazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 158
Product Link https://store.apolloscientific.co.uk/product/5-bromo-1h-benzimidazole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-1h-benzimidazole-2-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart