APO455830663 Apollo Scientific Ltd

4-Bromo-1,5-dimethyl-1H-indazole

  • MDL number MFCD12028620
  • Cas Number : 1159511-77-9
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-15-dimethyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-15-dimethyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart