APO455830666 Apollo Scientific Ltd

5-Bromo-1,6-dimethyl-1H-indazole

  • MDL number MFCD12028625
  • Cas Number : 1159511-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45219

Safety Information

signalword Warning
pcodes P260;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45219
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart