APO455830669 Apollo Scientific Ltd

4-Bromo-2,5-dimethyl-2H-indazole

  • MDL number MFCD12028631
  • Cas Number : 1159511-86-0
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2,5-dimethyl-2H-indazole
Inchi Code 1S/C9H9BrN2/c1-6-3-4-8-7(9(6)10)5-12(2)11-8/h3-5H,1-2H3
Inchi Key QHLAFXZSLHLHOT-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-25-dimethyl-2h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-25-dimethyl-2h-indazole
pictograms GHS07
hstatements H302;H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart