APO455830670 Apollo Scientific Ltd

5-Bromo-2,3-dimethyl-2H-indazole

  • MDL number MFCD12028634
  • Cas Number : 878064-16-5
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-23-dimethyl-2h-indazole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-23-dimethyl-2h-indazole
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart