APO455830671 Apollo Scientific Ltd

5-Bromo-2,4-dimethyl-2H-indazole

  • MDL number MFCD12028635
  • Cas Number : 1159511-89-3
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-dimethyl-2H-indazole
Inchi Code 1S/C9H9BrN2/c1-6-7-5-12(2)11-9(7)4-3-8(6)10/h3-5H,1-2H3
Inchi Key CJUCAYMTGYRYEH-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-dimethyl-2h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-dimethyl-2h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart