APO455830680 Apollo Scientific Ltd

2-Amino-6-purinethiol

  • MDL number MFCD00127785
  • Cas Number : 154-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 6-Thioguanin, 2-Amino-6-mercaptopurine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26832

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26832
pictograms GHS06;GHS08
hstatements H301;H341
Availability details for
Quantity
  • Availability
  • Add to Cart