APO455830681 Apollo Scientific Ltd

1,3-Dimethyl-1H-indazole-5-boronic acid

  • MDL number MFCD12028654
  • Cas Number : 1310404-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Borono-1,3-dimethyl-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/13-dimethyl-1h-indazole-5-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-dimethyl-1h-indazole-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart