APO455830682 Apollo Scientific Ltd

1,4-Dimethyl-1H-indazole-5-boronic acid

  • MDL number MFCD12028655
  • Cas Number : 1310405-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Borono-1,4-dimethyl-1H-indazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45268

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45268
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart