APO455830691 Apollo Scientific Ltd

Piperidin-3-one hydrochloride

  • MDL number MFCD06796739
  • Cas Number : 61644-00-6
  • Molecular Weight : 135.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Azacyclohexan-1-one hydrochloride, 3-Oxopiperidine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/piperidin-3-one-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/piperidin-3-one-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart